Learn More
Medchemexpress LLC OS-3-106 50mg | 1580000-17-4 | 450.60 g/mol | C25H30N4O2S | 50 MG
SDP
Catalog No. 5000228204
Encompass_Preferred
A selective dopamine D3 receptor (D3R) agonist used as a research reagent in neuropharmacology and addiction studies. The compound is blood-brain barrier penetrant and binds D3R with high affinity (Ki = 0.2 nM). It is supplied as a solid with high purity and requires cold storage for extended stability.
  • Selective D3 receptor agonist with Ki = 0.2 nM.
  • Blood-brain barrier penetrant.
  • Supplied as an off-white to light yellow solid.
  • High purity suitable for research applications.
  • Stable under recommended cold storage conditions.
Catalog No. 50-002-28204 Supplier Medchemexpress LLC Supplier No. HY11682050MG
A selective dopamine D3 receptor (D3R) agonist used as a research reagent in neuropharmacology and addiction studies. The compound is blood-brain barrier penetrant and binds D3R with high affinity (Ki = 0.2 nM). It is supplied as a solid with high purity and requires cold storage for extended stability.
  • Selective D3 receptor agonist with Ki = 0.2 nM.
  • Blood-brain barrier penetrant.
  • Supplied as an off-white to light yellow solid.
  • High purity suitable for research applications.
  • Stable under recommended cold storage conditions.