Learn More
Medchemexpress LLC OS-3-106 5mg | 1580000-17-4 | 450.60 | C25H30N4O2S | 5 MG
SDP
Catalog No. 5000227611
Encompass_Preferred
OS-3-106 is a potent, blood-brain barrier-penetrant, selective dopamine D3 receptor (D3R) agonist used as a research reagent in neuroscience and addiction studies. Molecular formula C25H30N4O2S; molecular weight 450.60; purity 97.03%.
  • Selective D3 receptor agonist with Ki = 0.2 nM.
  • Blood-brain barrier penetrant, suitable for CNS research applications.
  • High purity suitable for biological assays.
  • Solid form, typically off-white to light yellow.
  • Available in small research quantities, such as 5 mg.
Catalog No. 50-002-27611 Supplier Medchemexpress LLC Supplier No. HY1168205MG
OS-3-106 is a potent, blood-brain barrier-penetrant, selective dopamine D3 receptor (D3R) agonist used as a research reagent in neuroscience and addiction studies. Molecular formula C25H30N4O2S; molecular weight 450.60; purity 97.03%.
  • Selective D3 receptor agonist with Ki = 0.2 nM.
  • Blood-brain barrier penetrant, suitable for CNS research applications.
  • High purity suitable for biological assays.
  • Solid form, typically off-white to light yellow.
  • Available in small research quantities, such as 5 mg.