Learn More
MSC Rubbermaid Liquid Soap Dispenser, Counter-mounted, Touchless, 800/1600 mL, Polished Chrome
Catalog No. 502836802
Encompass
The Rubbermaid FG402073 One-Shot Liquid Soap Dispenser is a counter-mounted unit designed for sanitary and efficient soap dispensing. It utilizes touchless Smart Sensor™ technology to deliver uniform portions of soap, reducing overuse. Constructed from durable metal with a polished chrome finish, this dispenser offers programmable options for 800 mL and 1600 mL refills.
  • Delivers uniform portions of soap
  • Touchless, sanitary Smart Sensor™ technology
  • Requires four D-cell batteries for operation
  • Indicator light for battery replacement
  • Durable metal with a polished chrome finish
  • Closed dispensing system
  • Dispenses uniform liquid volumes to reduce overuse
  • Battery life can last 3 years or 120,000 cycles
  • Indicator lights for battery life and fill level
  • ADA compliant
Catalog No. 50-283-6802 Supplier MSC Supplier No. 57797656
The Rubbermaid FG402073 One-Shot Liquid Soap Dispenser is a counter-mounted unit designed for sanitary and efficient soap dispensing. It utilizes touchless Smart Sensor™ technology to deliver uniform portions of soap, reducing overuse. Constructed from durable metal with a polished chrome finish, this dispenser offers programmable options for 800 mL and 1600 mL refills.
  • Delivers uniform portions of soap
  • Touchless, sanitary Smart Sensor™ technology
  • Requires four D-cell batteries for operation
  • Indicator light for battery replacement
  • Durable metal with a polished chrome finish
  • Closed dispensing system
  • Dispenses uniform liquid volumes to reduce overuse
  • Battery life can last 3 years or 120,000 cycles
  • Indicator lights for battery life and fill level
  • ADA compliant