Learn More
Medchemexpress LLC Osavampator 10mg | 1358751-06-0 | 10 MG
SDP
Catalog No. 5000229800
Encompass_Preferred
Osavampator is a small-molecule positive allosteric modulator of AMPA-type glutamate receptors used in research on cognition and depressive disorders. Supplied for laboratory research use only. Molecular formula C19H23N3O3S; molecular weight 373.47 g·mol⁻¹.
  • Positive allosteric modulator of AMPA receptors that enhances glutamatergic signaling.
  • Provided as a 10 mg vial suitable for preparation of assay concentrations.
  • High purity appropriate for in vitro pharmacology and biochemical assays.
  • Recommended storage: sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not approved for human therapeutic use.
Catalog No. 50-002-29800 Supplier Medchemexpress LLC Supplier No. HY11586410MG
Osavampator is a small-molecule positive allosteric modulator of AMPA-type glutamate receptors used in research on cognition and depressive disorders. Supplied for laboratory research use only. Molecular formula C19H23N3O3S; molecular weight 373.47 g·mol⁻¹.
  • Positive allosteric modulator of AMPA receptors that enhances glutamatergic signaling.
  • Provided as a 10 mg vial suitable for preparation of assay concentrations.
  • High purity appropriate for in vitro pharmacology and biochemical assays.
  • Recommended storage: sealed, away from moisture and light; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not approved for human therapeutic use.