Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osborn Shoe Caps

Catalog No. 19041924 Shop All Osborn Scientific Products
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Protect the entire top of the foot
Protects tip of foot
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
19-041924 Protects tip of foot
19-041925 Protect the entire top of the foot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-041924 Supplier Osborn Supplier No. OG3601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

For temporary foot protection

 

  • Toe Cap
  • Slip easily over shoes and strap firmly around back of heel
  • 13.6kg (30 lbs.) impact resistance
  • Plastic material
  • Color: Yellow

 

Compliance

ANSI standard Z-41-1991

Specifications

Certifications/Compliance ANSI Z41-1991
Color Yellow
For Use With (Application) Protects tip of foot
Material Plastic
Toe Style Toe guard
Product Type Shoe caps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.