Learn More
Aviva Systems Biology Corp OSBP Antibody - C-terminal region (ARP60212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSBP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OSBP. Alternate names: OSBP1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-976