Learn More
Aviva Systems Biology Corp OSBPL11 Antibody - C-terminal region (ARP45658_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45658_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSBPL11. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human OSBPL11. Alternate names: FLJ13012, FLJ13164, ORP-11, ORP11, OSBP12, TCCCIA00292ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-213