Learn More
Aviva Systems Biology Corp OSBPL1A Antibody - middle region (ARP45653_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45653_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSBPL1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OSBPL1A. Alternate names: FLJ10217, ORP-1, ORP1, OSBPL1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53891