Learn More
Aviva Systems Biology Corp OSBPL8 Antibody - N-terminal region (ARP44452_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44452_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSBPL8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OSBPL8. Alternate names: DKFZp686A11164, MGC126578, MGC133203, MST120, MSTP120, ORP8, OSBP10ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58815