Learn More
Aviva Systems Biology Corp OSBPL9 Antibody - N-terminal region (ARP45659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSBPL9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human OSBPL9. Alternate names: FLJ12492, FLJ14629, FLJ14801, FLJ32055, FLJ34384, MGC15035, ORP9, ORP-9ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59094