Learn More
MSC PRO-SOURCE Fan Head, 24-in, 1/4 hp, 2-Speed Oscillating, 1 ea
Catalog No. 501919859
Encompass
This industrial circulation fan head is designed for efficient air movement in various industrial settings. It features a 24 in diameter and a 1/4 hp motor, providing reliable performance.
  • Two-speed operation
  • Oscillating fan head
  • Operates at 120V
  • Airflow of 5000 CFM at low speed
  • Made in Czechia
Catalog No. 50-191-9859 Supplier MSC Supplier No. 38762522
This industrial circulation fan head is designed for efficient air movement in various industrial settings. It features a 24 in diameter and a 1/4 hp motor, providing reliable performance.
  • Two-speed operation
  • Oscillating fan head
  • Operates at 120V
  • Airflow of 5000 CFM at low speed
  • Made in Czechia