Learn More
Aviva Systems Biology Corp OSCAR Antibody - C-terminal region (ARP63083_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63083_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OSCAR. It was validated on Western Blot. . Alternate names: MGC33613, PIGR3ApplicationWB. Species reactivity: Human
Catalog No. 50-287-866