Learn More
Apexbio Technology LLC Oseltamivir, 100mg Cas# 196618-13-0
SDP
Supplier:  Apexbio Technology LLC A3688100
Encompass_Preferred
Oseltamivir is an inhibitor of influenza neuraminidase [1].Oseltamivir is a prodrug that is converted by intestinal and/or hepatic esterases to the neuraminidase inhibitor molecule, oseltamivir carboxylate. Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8515