Learn More
Apexbio Technology LLC Oseltamivir 196618-13-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063170
Encompass_Preferred
Oseltamivir is a neuraminidase inhibitor targeting influenza virus replication It functions as a prodrug metabolized into oseltamivir carboxylate by host esterases This active metabolite binds to viral neuraminidase thereby blocking cleavage of terminal -Neu5Ac residues and subsequent viral progeny release from infected host cells Oseltamivir has been widely employed in investigative studies addressing antiviral efficacy resistance mechanisms and clinical outcomes in influenza infection research The compound is frequently used in vitro to characterize neuraminidase activity inhibition typically reporting IC50 values in the low nanomolar range (approximately 0 1-5 nM) depending on the influenza viral strain and assay conditions influencing viral replication and transmission
Catalog No. 50-000-63170 Supplier Apexbio Technology LLC Supplier No. A36885.1
Oseltamivir is a neuraminidase inhibitor targeting influenza virus replication It functions as a prodrug metabolized into oseltamivir carboxylate by host esterases This active metabolite binds to viral neuraminidase thereby blocking cleavage of terminal -Neu5Ac residues and subsequent viral progeny release from infected host cells Oseltamivir has been widely employed in investigative studies addressing antiviral efficacy resistance mechanisms and clinical outcomes in influenza infection research The compound is frequently used in vitro to characterize neuraminidase activity inhibition typically reporting IC50 values in the low nanomolar range (approximately 0 1-5 nM) depending on the influenza viral strain and assay conditions influencing viral replication and transmission