Learn More
Medchemexpress LLC Oseltamivir-acetate | 1191921-01-3 | 99.5% | 354.44 | 25 MG
SDP
Catalog No. 5000401096
Encompass_Preferred
Oseltamivir-acetate is an impurity of Oseltamivir. Oseltamivir is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B.
  • For research use only.
Catalog No. 50-004-01096 Supplier Medchemexpress LLC Supplier No. 25MGHY43575
Oseltamivir-acetate is an impurity of Oseltamivir. Oseltamivir is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B.
  • For research use only.