Learn More
Cayman Chemical Oseltamivir Acid, Purity: ≥95%, CAS: 187227-45-8, 10 mg
SDP
Catalog No. 502972722
Encompass_Preferred
The active metabolite of the antiviral prodrug oseltamivir; acts as an inhibitor of influenza neuraminidases A and B (IC50 values are 0.1 to 4.9 nM for both enzymes), in this way preventing virus budding and release
Catalog No. 50-297-2722 Supplier Cayman Chemical Supplier No. 1577910 MG
The active metabolite of the antiviral prodrug oseltamivir; acts as an inhibitor of influenza neuraminidases A and B (IC50 values are 0.1 to 4.9 nM for both enzymes), in this way preventing virus budding and release