Learn More
Medchemexpress LLC HY-13318 50mg , Oseltamivir (acid) GS 4071;Ro 64-0802;oseltamivir carboxylate CAS:187227-45-8 Purity:98%
SDP
Catalog No. 501493614
Encompass_Preferred
Medchemexpress, HY-13318 50mg Oseltamivir (acid) GS 4071;Ro 64-0802;oseltamivir carboxylate CAS:187227-45-8 Formula:C14H24N2O4 Influenza A and B Purity:98% Oseltamivir acid is an active metabolite of Oseltamivir, which is a potent and selective inhibitor of influenza A and B virus neuraminidases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3614 Supplier Medchemexpress LLC Supplier No. HY1331850MG
Medchemexpress, HY-13318 50mg Oseltamivir (acid) GS 4071;Ro 64-0802;oseltamivir carboxylate CAS:187227-45-8 Formula:C14H24N2O4 Influenza A and B Purity:98% Oseltamivir acid is an active metabolite of Oseltamivir, which is a potent and selective inhibitor of influenza A and B virus neuraminidases. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.