Learn More
Medchemexpress LLC Oseltamivir acid-d3 | 1242184-43-5 | MFCD08703074 | 98.4% | 287.37 g/mol | C14H21D3N2O4 | 5 MG
SDP
Catalog No. 5000262397
Encompass_Preferred
Oseltamivir acid-d3 is a deuterium-labeled isotope analogue of oseltamivir acid (the active carboxylate metabolite). Provided as a solid reference standard, it is intended for analytical use including method development, isotope-dilution quantification, and pharmacokinetic studies. Typical identifiers: CAS 1242184-43-5; formula C14H21D3N2O4; molecular weight 287.37 g/mol; purity ~98.4%.
  • Deuterium-labeled internal standard for LC-MS/MS quantification.
  • High chemical purity suitable for quantitative analysis.
  • Small solid format for convenient storage and handling.
  • Appropriate for method validation and pharmacokinetic assays.
Catalog No. 50-002-62397 Supplier Medchemexpress LLC Supplier No. HY13318S5MG
Oseltamivir acid-d3 is a deuterium-labeled isotope analogue of oseltamivir acid (the active carboxylate metabolite). Provided as a solid reference standard, it is intended for analytical use including method development, isotope-dilution quantification, and pharmacokinetic studies. Typical identifiers: CAS 1242184-43-5; formula C14H21D3N2O4; molecular weight 287.37 g/mol; purity ~98.4%.
  • Deuterium-labeled internal standard for LC-MS/MS quantification.
  • High chemical purity suitable for quantitative analysis.
  • Small solid format for convenient storage and handling.
  • Appropriate for method validation and pharmacokinetic assays.