Learn More
Medchemexpress LLC Oseltamivir acid (hydrochloride) | 1415963-60-8 | >98.0% | 320.81 g/mol | C14H25ClN2O4 | 5 MG
SDP
Catalog No. 5000262554
Encompass_Preferred
Oseltamivir acid (hydrochloride) is the active metabolite of the antiviral prodrug oseltamivir; it is a potent, selective influenza virus neuraminidase inhibitor supplied for research use as a reference standard and assay reagent.
  • Active metabolite used in antiviral research.
  • Potent and selective neuraminidase inhibitor.
  • Suitable as a reference standard for biochemical and pharmacology studies.
  • Available in small quantities for laboratory use.
  • Batch-specific certificate of analysis available for purity verification.
Catalog No. 50-002-62554 Supplier Medchemexpress LLC Supplier No. HY13318A5MG
Oseltamivir acid (hydrochloride) is the active metabolite of the antiviral prodrug oseltamivir; it is a potent, selective influenza virus neuraminidase inhibitor supplied for research use as a reference standard and assay reagent.
  • Active metabolite used in antiviral research.
  • Potent and selective neuraminidase inhibitor.
  • Suitable as a reference standard for biochemical and pharmacology studies.
  • Available in small quantities for laboratory use.
  • Batch-specific certificate of analysis available for purity verification.