Learn More
Medchemexpress LLC Oseltamivir-d3 phosphate | 2734919-89-0 | 413.42 | C16H28D3N2O8P | 5 MG
SDP
Catalog No. 5000284724
Encompass_Preferred
Oseltamivir-d3 Phosphate is the deuterium-labeled form of oseltamivir phosphate supplied for research use as a tracer and internal standard in quantitative analytical workflows such as NMR, GC-MS, and LC-MS. It is intended for scientific research and drug declaration purposes only and not for personal or clinical use.
  • Deuterium-labeled internal standard for analytical assays.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Provided in small, analytical-scale quantities for method development and calibration.
  • Product datasheet available for purity and product data details.
Catalog No. 50-002-84724 Supplier Medchemexpress LLC Supplier No. HYW6538775MG
Oseltamivir-d3 Phosphate is the deuterium-labeled form of oseltamivir phosphate supplied for research use as a tracer and internal standard in quantitative analytical workflows such as NMR, GC-MS, and LC-MS. It is intended for scientific research and drug declaration purposes only and not for personal or clinical use.
  • Deuterium-labeled internal standard for analytical assays.
  • Suitable for NMR, GC-MS, and LC-MS quantitative analysis.
  • Provided in small, analytical-scale quantities for method development and calibration.
  • Product datasheet available for purity and product data details.