Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oseltamivir-d5 | 1093851-63-8 | 98.5% | 317.44 g/mol | C16H23D5N2O4 | 5 MG
SDP

Catalog No. 5000262550
Encompass_Preferred

Oseltamivir-d5 is the deuterium-labeled analogue of oseltamivir supplied as a solid powder and used as an analytical reference and internal standard in mass spectrometry and pharmacokinetic studies. Molecular formula C16H23D5N2O4, molecular weight 317.44 g/mol, CAS 1093851-63-8. Typical packaging is a 5 mg vial and reported purity is ~98.49%.

  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High chemical purity (~98.5%) suitable for analytical use.
  • Solid powder form, facilitating accurate weighing and solution preparation.
  • Useful for pharmacokinetic and bioanalytical studies to distinguish analyte by mass.
  • Molecular weight 317.44 g/mol and formula C16H23D5N2O4 for reference.

Catalog No. 50-002-62550 Supplier Medchemexpress LLC Supplier No. HY13317S35MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oseltamivir-d5 is the deuterium-labeled analogue of oseltamivir supplied as a solid powder and used as an analytical reference and internal standard in mass spectrometry and pharmacokinetic studies. Molecular formula C16H23D5N2O4, molecular weight 317.44 g/mol, CAS 1093851-63-8. Typical packaging is a 5 mg vial and reported purity is ~98.49%.

  • Deuterium-labeled internal standard for mass spectrometry and quantitative analysis.
  • High chemical purity (~98.5%) suitable for analytical use.
  • Solid powder form, facilitating accurate weighing and solution preparation.
  • Useful for pharmacokinetic and bioanalytical studies to distinguish analyte by mass.
  • Molecular weight 317.44 g/mol and formula C16H23D5N2O4 for reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.