Learn More
Medchemexpress LLC Oseltamivir-d5 phosphate | 415.43 | C16H26D5N2O8P | 1 MG
SDP
Catalog No. 5000283921
Encompass_Preferred
Oseltamivir-d5 phosphate is a deuterium-labeled analogue of oseltamivir phosphate used as a stable isotope internal standard and tracer for quantitative analysis in biological and chemical assays. It is intended for research use only and is commonly used in LC-MS, GC-MS, and NMR workflows for accurate quantitation.
  • Deuterium-labeled internal standard for oseltamivir analyses.
  • Enables accurate quantitative analysis by LC-MS, GC-MS, and NMR.
  • Formula C16H26D5N2O8P; molecular weight 415.43 g/mol.
  • Supplied in small analytical quantities suitable for trace-level studies.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-83921 Supplier Medchemexpress LLC Supplier No. HYW6543011MG
Oseltamivir-d5 phosphate is a deuterium-labeled analogue of oseltamivir phosphate used as a stable isotope internal standard and tracer for quantitative analysis in biological and chemical assays. It is intended for research use only and is commonly used in LC-MS, GC-MS, and NMR workflows for accurate quantitation.
  • Deuterium-labeled internal standard for oseltamivir analyses.
  • Enables accurate quantitative analysis by LC-MS, GC-MS, and NMR.
  • Formula C16H26D5N2O8P; molecular weight 415.43 g/mol.
  • Supplied in small analytical quantities suitable for trace-level studies.
  • Intended for research use only; not for human or veterinary use.