Learn More
Selleck Chemical LLC Oseltamivir Phosphate S2597-1g
SDP
Catalog No. 503112548
Encompass_Preferred
Oseltamivir Phosphate(Tamiflu) is a potent and selective inhibitor of the neuraminidase that is essential for replication of influenza A and B viruses used to prevent influenza
Catalog No. 50-311-2548 Supplier Selleck Chemical LLC Supplier No. S25971G
Oseltamivir Phosphate(Tamiflu) is a potent and selective inhibitor of the neuraminidase that is essential for replication of influenza A and B viruses used to prevent influenza