Learn More
Medchemexpress LLC Oseltamivir phosphate | 204255-11-8 | 410.40 | 10 MM/1 ML
SDP
Catalog No. 5000384416
Encompass_Preferred
Oseltamivir phosphate (GS 4104) is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B. It is a prodrug that is readily absorbed from the gastrointestinal tract and converted predominantly by hepatic esterases to Oseltamivir carboxylate (OC), acting as a widely used anti-influenza sialidase inhibitor.
  • Neuraminidase inhibitor for influenza A and B
  • Prodrug form, converted to Oseltamivir carboxylate (OC)
  • Used for treatment and prophylaxis of influenza
Catalog No. 50-003-84416 Supplier Medchemexpress LLC Supplier No. HY1701610MM/1ML
Oseltamivir phosphate (GS 4104) is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B. It is a prodrug that is readily absorbed from the gastrointestinal tract and converted predominantly by hepatic esterases to Oseltamivir carboxylate (OC), acting as a widely used anti-influenza sialidase inhibitor.
  • Neuraminidase inhibitor for influenza A and B
  • Prodrug form, converted to Oseltamivir carboxylate (OC)
  • Used for treatment and prophylaxis of influenza