Learn More
AdipoGen Oseltamivir . phosphate salt (250 mg)
SDP
Supplier:  AdipoGen AGCR13714M250
Encompass_Preferred
Chemical. CAS: 204255-11-8. Formula: C16H28N2O4 . H3PO4. MW: 312.4 . 98.0. Oseltamivir phosphate is an influenza viral neuraminidase inhibitor. It is an antiviral prodrug targeted against the influenza viruses. Once hydrolyzed in the liver to its active metabolite, oseltamivir carboxylate, it can competitively inhibit viral neuraminidase (IC50s=0.1-4.9nM for influenza neuraminidases A and B). The enzyme cleaves the sialic acid which is found on glycoproteins on the surface of human cells that helps new virions to exit the cell. Thus oseltamivir is blocking the release of new viral particles from a host cell. Oseltamivir phosphate is used clinically to treat influenza A and influenza B, and to prevent flu after exposure. It potentially could be used to reduce the spread of the SARS-CoV2 and counteract against the transmission of COVID-19.
Catalog No. 50-205-4099