Learn More
Apexbio Technology LLC Oseltamivir phosphate, 500mg Cas# 204255-11-8
SDP
Supplier:  Apexbio Technology LLC B1803500
Encompass_Preferred
Oseltamivir phosphate (GS-4104) is a competitive neuraminidase inhibitor, which is an antiviral drug. 204255-11-8. MFCD17676100 Oseltamivir phosphate (GS-4104) is a competitive neuraminidase inhibitor, which is an antiviral drug. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8649