Learn More
Apexbio Technology LLC OSELTAMIVIR-PHOSPHATE-D5-1MG
SDP
Catalog No. 5000570273
Encompass_Preferred
Oseltamivir phosphate-d5 (CAS No 204255-11-8) is an isotopically labeled compound used extensively in biomedical research particularly within virology and pharmacokinetic studies It serves as a stable isotope-labeled internal standard for the quantification and analysis of oseltamivir an antiviral medication used to treat and prevent influenza A and B The mechanism of action involves inhibiting the neuraminidase enzyme which is crucial for viral replication and release from host cells In vitro activity of oseltamivir phosphate-d5 generally falls within the nanomolar range indicating high potency in enzyme assays Researchers often employ this compound in cell models and animal studies to better understand drug metabolism and conduct drug screening processes Experimental concentrations typically depend on the specific research design allowing for a broad application across various studies
Catalog No. 50-005-70273 Supplier Apexbio Technology LLC Supplier No. SL10651
Oseltamivir phosphate-d5 (CAS No 204255-11-8) is an isotopically labeled compound used extensively in biomedical research particularly within virology and pharmacokinetic studies It serves as a stable isotope-labeled internal standard for the quantification and analysis of oseltamivir an antiviral medication used to treat and prevent influenza A and B The mechanism of action involves inhibiting the neuraminidase enzyme which is crucial for viral replication and release from host cells In vitro activity of oseltamivir phosphate-d5 generally falls within the nanomolar range indicating high potency in enzyme assays Researchers often employ this compound in cell models and animal studies to better understand drug metabolism and conduct drug screening processes Experimental concentrations typically depend on the specific research design allowing for a broad application across various studies