Learn More
Cayman Chemical OseltamIvIrphosphate 100mg
SDP
Catalog No. 502973071
Encompass_Preferred
An antiviral prodrug targeted against the influenza viruses; once hydrolyzed to its active metabolite, it can competitively inhibit viral neuraminidase (IC50s = 0.1-4.9 nM for influenza neuraminidases A and B)
Catalog No. 50-297-3071 Supplier Cayman Chemical Supplier No. 16070100 MG
An antiviral prodrug targeted against the influenza viruses; once hydrolyzed to its active metabolite, it can competitively inhibit viral neuraminidase (IC50s = 0.1-4.9 nM for influenza neuraminidases A and B)