Learn More
MSC OSG Plug Thread Gage, 5/16-18 UNC, 2B Class, HSS, 1 ea
Catalog No. 5000514817
Encompass
This double-ended plug thread gage is designed for the precise inspection of pitch diameter and pitch accuracy of internal threads. Constructed from high-speed steel and hardened to 64 HRC, this instrument provides high durability and reliable performance in industrial thread checking applications.
  • Thread size: 5/16-18 UNC
  • Class of fit: 2B
  • Gage type: Go and No Go, double end
  • Material: High-speed steel
  • Tolerance: Class X per ANSI B1.2
  • Coating: Bright
  • Hardness: 64 HRC
Catalog No. 50-005-14817 Supplier MSC Supplier No. 55439947
This double-ended plug thread gage is designed for the precise inspection of pitch diameter and pitch accuracy of internal threads. Constructed from high-speed steel and hardened to 64 HRC, this instrument provides high durability and reliable performance in industrial thread checking applications.
  • Thread size: 5/16-18 UNC
  • Class of fit: 2B
  • Gage type: Go and No Go, double end
  • Material: High-speed steel
  • Tolerance: Class X per ANSI B1.2
  • Coating: Bright
  • Hardness: 64 HRC