Learn More
MSC OSG Plug Thread Gage, #10-32 Thread, 2B Class, Double End
Catalog No. 5000515022
Encompass
This Plug Thread Gage is designed for precision measurement of #10-32 threads. It features a double-end design with both "Go" and "No Go" ends, ensuring accurate validation of internal threads. The gage includes a handle for ease of use and is constructed from High-Speed Steel for durability. It has a 2B classification and includes NIST Traceability Certification, conforming to NIST standards.
  • Double-end design with "Go" and "No Go" functions
  • Measures #10-32 threads
  • Made from high-speed steel
  • 2B classification
  • Go pitch diameter of 0.1697 inch
  • No Go pitch diameter of 0.1736 inch
  • Handle is included
  • Includes NIST traceability certification
  • Conforms to NIST standards
Catalog No. 50-005-15022 Supplier MSC Supplier No. 39361001
This Plug Thread Gage is designed for precision measurement of #10-32 threads. It features a double-end design with both "Go" and "No Go" ends, ensuring accurate validation of internal threads. The gage includes a handle for ease of use and is constructed from High-Speed Steel for durability. It has a 2B classification and includes NIST Traceability Certification, conforming to NIST standards.
  • Double-end design with "Go" and "No Go" functions
  • Measures #10-32 threads
  • Made from high-speed steel
  • 2B classification
  • Go pitch diameter of 0.1697 inch
  • No Go pitch diameter of 0.1736 inch
  • Handle is included
  • Includes NIST traceability certification
  • Conforms to NIST standards