Learn More
MSC OSG Plug Thread Gage, M3.5 x 0.6, 6H Class, Double End, Go/No Go
Catalog No. 5000529043
Encompass
Metric plug thread gages are designed to inspect the pitch diameter and pitch accuracy of internal threads. These gages are manufactured to Class X tolerances per ANSI B1.16M, ensuring precise measurements. Constructed from ground premium HSS, they are hardened to 64 HRC for superior wear resistance.
  • Available with short form or long form certification
  • Same day shipping for short form certificates
  • Comes in standard 6H class of fit
  • Hardened to 64 HRC for durability
  • Provides superior wear resistance
  • Made from premium high speed steel
Catalog No. 50-005-29043 Supplier MSC Supplier No. 55439897
Metric plug thread gages are designed to inspect the pitch diameter and pitch accuracy of internal threads. These gages are manufactured to Class X tolerances per ANSI B1.16M, ensuring precise measurements. Constructed from ground premium HSS, they are hardened to 64 HRC for superior wear resistance.
  • Available with short form or long form certification
  • Same day shipping for short form certificates
  • Comes in standard 6H class of fit
  • Hardened to 64 HRC for durability
  • Provides superior wear resistance
  • Made from premium high speed steel