Learn More
MSC OSG Thread Gage, M24 x 1.5, 6H Class, Go & No Go, HSS
Catalog No. 5000527668
Encompass
These metric plug thread gages are designed to inspect the pitch diameter and accuracy of internal threads. Manufactured to Class X tolerances per ANSI B1.16M, these Go and No-Go gages are made from premium high-speed steel and hardened to 64 Rc for superior wear resistance.
  • Double-end design for efficient measurement.
  • Manufactured to Class X tolerances per ANSI B1.16M.
  • Available with short or long form certificates of conformance.
  • Hardened to 64 HRC for superior wear resistance.
Catalog No. 50-005-27668 Supplier MSC Supplier No. 55440390
These metric plug thread gages are designed to inspect the pitch diameter and accuracy of internal threads. Manufactured to Class X tolerances per ANSI B1.16M, these Go and No-Go gages are made from premium high-speed steel and hardened to 64 Rc for superior wear resistance.
  • Double-end design for efficient measurement.
  • Manufactured to Class X tolerances per ANSI B1.16M.
  • Available with short or long form certificates of conformance.
  • Hardened to 64 HRC for superior wear resistance.