Learn More
MSC OSG Thread Limit Gage, #6-32 UNC, Class 2B, Go/No Go
Catalog No. 5000530281
Encompass
This thread limit gage is designed to inspect the pitch diameter and pitch accuracy of internal threads. Manufactured to Class X tolerances per ANSI B1.2, it features Go and No-Go gages that are precision-ground from high speed steel and hardened to 64 HRC for superior wear resistance.
  • Available from stock with short form or long form certification
  • Hardened to 64 HRC to provide superior wear resistance
  • Made from high speed steel
  • Available in standard 2B class of fit
  • Short form certificates of conformance are available for no charge
  • Long form certificates are available upon request
Catalog No. 50-005-30281 Supplier MSC Supplier No. 55440036
This thread limit gage is designed to inspect the pitch diameter and pitch accuracy of internal threads. Manufactured to Class X tolerances per ANSI B1.2, it features Go and No-Go gages that are precision-ground from high speed steel and hardened to 64 HRC for superior wear resistance.
  • Available from stock with short form or long form certification
  • Hardened to 64 HRC to provide superior wear resistance
  • Made from high speed steel
  • Available in standard 2B class of fit
  • Short form certificates of conformance are available for no charge
  • Long form certificates are available upon request