Learn More
MSC OSG Thread Limit Gage, M6 X 0.75, 6H Class, HSS, Go/No-Go
Catalog No. 5000530187
Encompass
This thread limit gage is designed for inspecting the pitch diameter and pitch accuracy of internal threads with an M6 X 0.75 specification. Constructed from premium High-Speed Steel (HSS), it features a double-ended Go/No-Go design for precise measurements. Manufactured to Class X tolerances and hardened to 64 HrC, it offers superior wear resistance for demanding industrial applications.
  • Inspects pitch diameter and accuracy of internal threads.
  • Manufactured to Class X tolerances per ANSI B1.16M.
  • Go and No-Go design made from premium HSS and hardened to 64 HrC for superior wear resistance.
  • Available with short form or long form certification.
  • Standard 6H class of fit.
Catalog No. 50-005-30187 Supplier MSC Supplier No. 55439863
Add to Cart
Add to Cart
This thread limit gage is designed for inspecting the pitch diameter and pitch accuracy of internal threads with an M6 X 0.75 specification. Constructed from premium High-Speed Steel (HSS), it features a double-ended Go/No-Go design for precise measurements. Manufactured to Class X tolerances and hardened to 64 HrC, it offers superior wear resistance for demanding industrial applications.
  • Inspects pitch diameter and accuracy of internal threads.
  • Manufactured to Class X tolerances per ANSI B1.16M.
  • Go and No-Go design made from premium HSS and hardened to 64 HrC for superior wear resistance.
  • Available with short form or long form certification.
  • Standard 6H class of fit.