Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs OSHA Notice Safety Sign: Safety Glasses With Side Shields Required In This Area

Catalog No. 19805239 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-805-239 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-239 Supplier Accuform Signs Supplier No. MPPE830XT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Notifies any persons entering area that a specific type of eye protection must be worn.

  • Avoid side impact eye injury by alerting employees to wear safety glasses with side protection
  • Bold black text with large blue header against white background
  • Traditional OSHA-style signs, are the most widely recognized design format in the industry
  • Notice header indicates information considered to be important but not hazard-related
  • Semi-rigid .060 in. thick polycarbonate with a protective coating
  • Protected by UV protective coating
  • Resists abrasion, humidity, and chemicals
  • Service temperature range: -40° to 251°F

Specifications

Material Plastic
Style OSHA
Resistant To Abrasion, Humidity, Chemicals
Description Dura-Plastic
Height (English) 10 in.
Max. Temperature (English) 251°F
Min. Temperature (English) -40°F
Thickness 0.060 in.
Width (English) 14 in.
Temperature Range (English) -40°F to 251°F
Languages English
Displayed Text Safety Glasses With Side This Area
Color White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.