Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oshcl(Co)[Kappa3-Pych2Nhc2H4Nhptbu2]

Catalog No. 808008500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-800-8500MG 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 80-800-8500MG Supplier Sigma Aldrich Supplier No. 808008500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

OsHCl(CO)[+a3-PyCH2NHC2H4NHPtBu2] is a robust and highly efficient precatalyst for coupling amines with methanol and ethanol to afford formamides and acetamides. It possesses excellent carbonyl selectivity with an unique activity for acceptorless dehydrogenative coupling of alcohols to esters and amides.

Specifications

CAS 1499-55-4
Melting Point 182°C
Molecular Formula C6H11NO4
Linear Formula CH3OCOCH2CH2CH(NH2)COOH
MDL Number MFCD00002632
Quantity 25 g
Molecular Weight (g/mol) 161.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.