Learn More
Apexbio Technology LLC OSI-930 728033-96-3 5mg
SDP
Catalog No. 5000096094
Encompass_Preferred
OSI-930 (CAS 728033-96-3) is a small molecule inhibitor targeting multiple receptor tyrosine kinases including Flt1 (IC 8 nM) KDR/VEGFR2 (IC 9 nM) CSF-1R (IC 15 nM) Lck c-Raf and Kit (IC 80 nM) In cellular studies OSI-930 inhibits proliferation and induces apoptosis in HMC-1 cells whose survival is dependent on Kit signaling while exhibiting minimal effects on COLO-205 cells under standard conditions Additionally OSI-930 inactivates cytochrome P450 3A4 in a time- and concentration-dependent manner and alters its spectral properties These characteristics make OSI-930 a valuable tool compound for investigating receptor tyrosine kinase signaling pathways and related cellular processes in cancer research
Catalog No. 50-000-96094 Supplier Apexbio Technology LLC Supplier No. A28385
OSI-930 (CAS 728033-96-3) is a small molecule inhibitor targeting multiple receptor tyrosine kinases including Flt1 (IC 8 nM) KDR/VEGFR2 (IC 9 nM) CSF-1R (IC 15 nM) Lck c-Raf and Kit (IC 80 nM) In cellular studies OSI-930 inhibits proliferation and induces apoptosis in HMC-1 cells whose survival is dependent on Kit signaling while exhibiting minimal effects on COLO-205 cells under standard conditions Additionally OSI-930 inactivates cytochrome P450 3A4 in a time- and concentration-dependent manner and alters its spectral properties These characteristics make OSI-930 a valuable tool compound for investigating receptor tyrosine kinase signaling pathways and related cellular processes in cancer research