Learn More
Medchemexpress LLC OSI-930 | 728033-96-3 | MFCD11100358 | 98.3% | 443.44 | C22H16F3N3O2S | 10 MG
SDP
Catalog No. 5000247436
Encompass_Preferred
OSI-930 is a small-molecule tyrosine kinase inhibitor selective for c-Kit, KDR (VEGFR2), and CSF-1R (c-Fms), provided as a solid for research use. It exhibits in vitro potency with reported IC50s of ~80 nM (Kit), 9 nM (KDR), and 15 nM (CSF-1R) and has shown antitumor activity in preclinical models.
  • Selective inhibition of c-Kit, KDR, and CSF-1R.
  • Low nanomolar potency against VEGFR2 (KDR).
  • Useful for kinase inhibition and cancer biology research.
  • High listed purity (≈98.3%).
  • Supplied as a solid in small-scale quantities suitable for laboratory studies.
Catalog No. 50-002-47436 Supplier Medchemexpress LLC Supplier No. HY1020410MG
OSI-930 is a small-molecule tyrosine kinase inhibitor selective for c-Kit, KDR (VEGFR2), and CSF-1R (c-Fms), provided as a solid for research use. It exhibits in vitro potency with reported IC50s of ~80 nM (Kit), 9 nM (KDR), and 15 nM (CSF-1R) and has shown antitumor activity in preclinical models.
  • Selective inhibition of c-Kit, KDR, and CSF-1R.
  • Low nanomolar potency against VEGFR2 (KDR).
  • Useful for kinase inhibition and cancer biology research.
  • High listed purity (≈98.3%).
  • Supplied as a solid in small-scale quantities suitable for laboratory studies.