Learn More
Medchemexpress LLC Osi-930 | 728033-96-3 | MFCD11100358 | 98.3% | 443.44 g/mol | C22H16F3N3O2S | 1 MG
SDP
Catalog No. 5000247597
Encompass_Preferred
OSI-930 is a small-molecule tyrosine kinase inhibitor selective for c-Kit, KDR (VEGFR2), and CSF-1R (c-Fms). It is used in preclinical research to probe kinase signaling, inhibit angiogenesis, and study antitumor effects; the compound is supplied in solid and solution formats for laboratory use.
  • Selective inhibitor of c-Kit, KDR (VEGFR2), and CSF-1R.
  • Potent biochemical activity with low-nanomolar IC50 values (example: KDR ≈9 nM).
  • Supplied in solid and solution forms, available in small research pack sizes.
  • Molecular weight 443.44 g/mol; formula C22H16F3N3O2S.
  • High purity (≈98% by RP-HPLC), suitable for preclinical assays.
Catalog No. 50-002-47597 Supplier Medchemexpress LLC Supplier No. HY102041MG
OSI-930 is a small-molecule tyrosine kinase inhibitor selective for c-Kit, KDR (VEGFR2), and CSF-1R (c-Fms). It is used in preclinical research to probe kinase signaling, inhibit angiogenesis, and study antitumor effects; the compound is supplied in solid and solution formats for laboratory use.
  • Selective inhibitor of c-Kit, KDR (VEGFR2), and CSF-1R.
  • Potent biochemical activity with low-nanomolar IC50 values (example: KDR ≈9 nM).
  • Supplied in solid and solution forms, available in small research pack sizes.
  • Molecular weight 443.44 g/mol; formula C22H16F3N3O2S.
  • High purity (≈98% by RP-HPLC), suitable for preclinical assays.