Learn More
Selleck Chemical LLC OSI-930 5mg 728033-96-3
SDP
Supplier:  Selleck Chemical LLC S12205MG
Encompass_Preferred
OSI-930 is a potent inhibitor of Kit (c-Kit), KDR and CSF-1R with IC50 of 80 nM, 9 nM and 15 nM, respectively; also potent to Flt-1, c-Raf and Lck and low activity against PDGFR?/?, Flt-3 and Abl. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-690-6