Learn More
Cayman Chemical OSILODROSTAT 10MG
SDP
Catalog No. 502946831
Encompass_Preferred
An aldosterone synthase inhibitor; selective for human aldosterone synthase over rat aldosterone synthase (IC50 = 160 nM); inhibits increases in plasma aldosterone levels induced by angiotensin II or ACTH in rats (ED50s = 0.6 and 1.1 mg/kg, respectively); reduces mean arterial pressure and prolongs survival in rats expressing human renin and angiotensinogen, a double-transgenic model of angiotensin II-dependent hypertension, in a dose-dependent manner
Catalog No. 50-294-6831 Supplier Cayman Chemical Supplier No. 3392810 MG
An aldosterone synthase inhibitor; selective for human aldosterone synthase over rat aldosterone synthase (IC50 = 160 nM); inhibits increases in plasma aldosterone levels induced by angiotensin II or ACTH in rats (ED50s = 0.6 and 1.1 mg/kg, respectively); reduces mean arterial pressure and prolongs survival in rats expressing human renin and angiotensinogen, a double-transgenic model of angiotensin II-dependent hypertension, in a dose-dependent manner