Learn More
Medchemexpress LLC HY-16276 2mg Medchemexpress, Osilodrostat CAS:928134-65-0 Purity:>98%
SDP
Catalog No. 501932766
Encompass_Preferred
Medchemexpress, HY-16276 2mg Osilodrostat CAS:928134-65-0 Osilodrostat (LCI699) is a potent inhibitor of human 11β-hydroxylase and aldosterone synthase with IC 50 values of 2.5 and 0.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2766 Supplier Medchemexpress LLC Supplier No. HY162762MG
Medchemexpress, HY-16276 2mg Osilodrostat CAS:928134-65-0 Osilodrostat (LCI699) is a potent inhibitor of human 11β-hydroxylase and aldosterone synthase with IC 50 values of 2.5 and 0.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.