Learn More
Cayman Chemical Osilodrostat, Purity: ≥98%, CAS: 928134-65-0, 50 mg
SDP
Catalog No. 502946833
Encompass_Preferred
An aldosterone synthase inhibitor; selective for human aldosterone synthase over rat aldosterone synthase (IC50 = 160 nM); inhibits increases in plasma aldosterone levels induced by angiotensin II or ACTH in rats (ED50s = 0.6 and 1.1 mg/kg, respectively); reduces mean arterial pressure and prolongs survival in rats expressing human renin and angiotensinogen, a double-transgenic model of angiotensin II-dependent hypertension, in a dose-dependent manner
Catalog No. 50-294-6833 Supplier Cayman Chemical Supplier No. 3392850 MG
An aldosterone synthase inhibitor; selective for human aldosterone synthase over rat aldosterone synthase (IC50 = 160 nM); inhibits increases in plasma aldosterone levels induced by angiotensin II or ACTH in rats (ED50s = 0.6 and 1.1 mg/kg, respectively); reduces mean arterial pressure and prolongs survival in rats expressing human renin and angiotensinogen, a double-transgenic model of angiotensin II-dependent hypertension, in a dose-dependent manner