Learn More
Medchemexpress LLC Osilodrostat phosphate (LCI699 phosphate) | 1315449-72-9 | 98.8% | 325.23 | 50 MG
SDP
Catalog No. 5000394973
Encompass_Preferred
Osilodrostat phosphate is a potent, orally active 11β-hydroxylase (CYP11B1) inhibitor with an IC50 value of 35 nM. It is also a potent, orally active aldosterone synthase (CYP11B2) inhibitor with IC50 values of 0.7 nM and 160 nM for human and rat aldosterone synthase, respectively. This compound inhibits aldosterone and corticosterone synthesis and has blood pressure-lowering ability. It can be used for research related to Cushing syndrome (CS).
  • Potent and orally active inhibitor of 11β-hydroxylase.
  • Potent and orally active inhibitor of aldosterone synthase.
  • Inhibits aldosterone and corticosterone synthesis.
  • Demonstrates blood pressure-lowering ability.
  • Useful for research of Cushing syndrome.
Catalog No. 50-003-94973 Supplier Medchemexpress LLC Supplier No. HY16276A50MG
Osilodrostat phosphate is a potent, orally active 11β-hydroxylase (CYP11B1) inhibitor with an IC50 value of 35 nM. It is also a potent, orally active aldosterone synthase (CYP11B2) inhibitor with IC50 values of 0.7 nM and 160 nM for human and rat aldosterone synthase, respectively. This compound inhibits aldosterone and corticosterone synthesis and has blood pressure-lowering ability. It can be used for research related to Cushing syndrome (CS).
  • Potent and orally active inhibitor of 11β-hydroxylase.
  • Potent and orally active inhibitor of aldosterone synthase.
  • Inhibits aldosterone and corticosterone synthesis.
  • Demonstrates blood pressure-lowering ability.
  • Useful for research of Cushing syndrome.