Learn More
Medchemexpress LLC Osimertinib-d6 | 1638281-44-3 | MFCD00056410 | 99.6% | 505.64 g/mol | C28H27D6N7O2 | 10 MG
SDP
Catalog No. 5000288472
Encompass_Preferred
Osimertinib-d6 is a deuterium-labeled analogue of osimertinib supplied as an analytical standard for research and quantitative mass spectrometry applications. It provides a defined mass shift relative to the parent drug that enables accurate internal standardization in LC-MS and bioanalytical assays.
  • Deuterium-labeled internal standard for quantitative LC-MS and LC-MS/MS assays.
  • Provides a mass-shifted analogue of osimertinib for accurate quantification.
  • High purity (~99.6%), supporting reliable analytical results.
  • Supplied as a solid with recommended storage protected from light at 4°C; in solution store -80°C (6 months) or -20°C (1 month).
  • DMSO stock preparation: 125 mg/mL (247.21 mM) with warming and sonication as needed.
Catalog No. 50-002-88472 Supplier Medchemexpress LLC Supplier No. HY15772S10MG
Osimertinib-d6 is a deuterium-labeled analogue of osimertinib supplied as an analytical standard for research and quantitative mass spectrometry applications. It provides a defined mass shift relative to the parent drug that enables accurate internal standardization in LC-MS and bioanalytical assays.
  • Deuterium-labeled internal standard for quantitative LC-MS and LC-MS/MS assays.
  • Provides a mass-shifted analogue of osimertinib for accurate quantification.
  • High purity (~99.6%), supporting reliable analytical results.
  • Supplied as a solid with recommended storage protected from light at 4°C; in solution store -80°C (6 months) or -20°C (1 month).
  • DMSO stock preparation: 125 mg/mL (247.21 mM) with warming and sonication as needed.