Learn More
Medchemexpress LLC HY-79077 5mg Medchemexpress, Osimertinib (dimesylate) CAS:2070014-82-1 Purity:>98%
SDP
Catalog No. 502082293
Encompass_Preferred
Medchemexpress, HY-79077 5mg Osimertinib (dimesylate) CAS:2070014-82-1 Osimertinib dimesylate (AZD-9291 dimesylate) is an irreversible and mutant selective EGFR inhibitor with IC50s of 12 and 1 nM against EGFRL858R and EGFRL858R/T790M, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2293 Supplier Medchemexpress LLC Supplier No. HY790775MG
Medchemexpress, HY-79077 5mg Osimertinib (dimesylate) CAS:2070014-82-1 Osimertinib dimesylate (AZD-9291 dimesylate) is an irreversible and mutant selective EGFR inhibitor with IC50s of 12 and 1 nM against EGFRL858R and EGFRL858R/T790M, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.