Learn More
Medchemexpress LLC HY-15772A 10mg Medchemexpress, Osimertinib mesylate CAS:1421373-66-1 Purity:>98%
SDP
Catalog No. 502029170
Encompass_Preferred
Medchemexpress, HY-15772A 10mg Osimertinib mesylate CAS:1421373-66-1 Osimertinib mesylate (AZD-9291 mesylate) is an irreversible and mutant selective EGFR inhibitor with IC50s of 12 and 1 nM against EGFRL858R and EGFRL858R/T790M, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9170 Supplier Medchemexpress LLC Supplier No. HY15772A10MG
Medchemexpress, HY-15772A 10mg Osimertinib mesylate CAS:1421373-66-1 Osimertinib mesylate (AZD-9291 mesylate) is an irreversible and mutant selective EGFR inhibitor with IC50s of 12 and 1 nM against EGFRL858R and EGFRL858R/T790M, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.