Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Osimertinib mesylate | 1421373-66-1 | MFCD28137994 | 99.9% | 595.71 | C29H37N7O5S | 10 MG
SDP

Catalog No. 5000288187
Encompass_Preferred

Osimertinib mesylate is the mesylate salt of osimertinib supplied as a high-purity solid intended for use as an analytical reference and research reagent. The material is provided with a certificate of analysis and safety data sheet for laboratory use and quality verification.

  • High purity: 99.93% suitable for analytical reference.
  • Solid form: stable powder for easy handling and storage.
  • Comprehensive documentation: COA and SDS available for quality and safety verification.
  • Multiple pack sizes: available in small to larger quantities to support different study scales.
  • Chemical identifiers provided: CAS number and MFCD number available for unambiguous identification.

Catalog No. 50-002-88187 Supplier Medchemexpress LLC Supplier No. HY15772AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Osimertinib mesylate is the mesylate salt of osimertinib supplied as a high-purity solid intended for use as an analytical reference and research reagent. The material is provided with a certificate of analysis and safety data sheet for laboratory use and quality verification.

  • High purity: 99.93% suitable for analytical reference.
  • Solid form: stable powder for easy handling and storage.
  • Comprehensive documentation: COA and SDS available for quality and safety verification.
  • Multiple pack sizes: available in small to larger quantities to support different study scales.
  • Chemical identifiers provided: CAS number and MFCD number available for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.