Learn More
Medchemexpress LLC HY-15772A 500mg , Osimertinib mesylate CAS:1421373-66-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15772A500MG
Encompass_Preferred
HY-15772A 500mg Osimertinib mesylate CAS: 1421373-66-1 Osimertinib mesylate (AZD9291 mesylate) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M. Osimertinib overcomes T790M-mediated resistance to EGFR inhibitors in lung cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0944