Learn More
Medchemexpress LLC HY-P7052 10ug Medchemexpress, OSM (227a.a.), Human CAS: Purity:>98%
SDP
Catalog No. 501934298
Encompass_Preferred
Medchemexpress, HY-P7052 10ug OSM (227a.a.), Human CAS: Recombinant Human Oncostatin-M, 209a.a. is a polypeptide chain containing 209 amino acids. Oncostatin M is a cytokine belonging to the IL-6 family that has multiple functions in hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, heart remodeling, nociception, inflammation and metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4298 Supplier Medchemexpress LLC Supplier No. HYP705210UG
Medchemexpress, HY-P7052 10ug OSM (227a.a.), Human CAS: Recombinant Human Oncostatin-M, 209a.a. is a polypeptide chain containing 209 amino acids. Oncostatin M is a cytokine belonging to the IL-6 family that has multiple functions in hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, heart remodeling, nociception, inflammation and metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.